Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033174.1 | NP_001028346.1 | 8 - 9 | 1646 | 133 | |
GenBank mRNA | AK132663.1 | - | 6 - 7 | 926 | 133 | |
AK220301.1 | - | 5 - 6 | 684 | 133 | ||
BC094342.1 | - | 2 - 3 | 267 | 133 | ||
BC107327.1 | - | 8 - 9 | 1380 | 133 | ||
BC107328.2 | - | 8 - 9 | 1380 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |