Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021555.2 | NP_067530.2 | 1 - 2 | 676 | 82 | |
GenBank mRNA | AB041552.1 | - | 1 - 2 | 676 | 82 | |
AK085815.1 | - | 1 - 2 | 670 | 82 | ||
AK194217.1 | - | 1 - 2 | 38 | 82 | ||
BC061251.1 | - | 1 - 2 | 640 | 30249656 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |