Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021555.2 | NP_067530.2 | 2 - 3 | 767 | 99 | |
GenBank mRNA | AB041552.1 | - | 2 - 3 | 767 | 99 | |
AK085815.1 | - | 2 - 3 | 761 | 99 | ||
BC061251.1 | - | 2 - 3 | 731 | 30249656 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |