Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700027287.mm.1 |
NM_001163662.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027287.mm.1 |
NM_001163662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027287.mm.1 |
NM_008749.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027287.mm.1 |
NM_008749.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027287.mm.1 |
TC0700003009.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027285.mm.1 |
NM_001163662.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027285.mm.1 |
NM_001163662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027285.mm.1 |
NM_008749.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027285.mm.1 |
NM_008749.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027285.mm.1 |
TC0700003009.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027290.mm.1 |
NM_001163662.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027290.mm.1 |
NM_001163662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027290.mm.1 |
NM_008749.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027290.mm.1 |
NM_008749.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027290.mm.1 |
TC0700003009.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700027295.mm.1 |
NM_001163662.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027295.mm.1 |
NM_001163662.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027295.mm.1 |
NM_008749.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027295.mm.1 |
NM_008749.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700027295.mm.1 |
TC0700003009.mm.1 |
MTA TC |