Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175130.4 | NP_780339.2 | 21 - 22 | 3395 | 59 | |
GenBank mRNA | AB112657.1 | - | 6 - 7 | 952 | 59 | |
AB112658.1 | - | 6 - 7 | 1150 | 59 | ||
AJ575814.1 | - | 21 - 22 | 3374 | 59 | ||
BC046472.1 | - | 11 - 12 | 2067 | 59 | ||
BC049165.1 | - | 19 - 20 | 3026 | 59 | ||
BC049993.1 | - | 7 - 8 | 1217 | 59 | ||
BC058632.1 | - | 14 - 15 | 2303 | 59 | ||
BC096475.1 | - | 21 - 22 | 3371 | 4225167 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |