Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tulp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF105712.1 | - | 6 - 7 | 575 | 102 | |
AK054304.1 | - | 3 - 4 | 533 | 102 | ||
AK077200.1 | - | 6 - 7 | 584 | 102 | ||
AK077211.1 | - | 3 - 4 | 381 | 102 | ||
AK161165.1 | - | 3 - 4 | 532 | 102 | ||
BB617192.1 | - | 6 - 7 | 584 | 102 | ||
BC089545.1 | - | 3 - 4 | 380 | 6770730 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |