Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164173.1 | NP_001157645.1 | 2 - 3 | 284 | 71 | |
NM_053193.2 | NP_444423.1 | 2 - 3 | 284 | 71 | ||
GenBank mRNA | AF322193.1 | - | 2 - 3 | 255 | 71 | |
AK155648.1 | - | 2 - 3 | 225 | 71 | ||
AK155806.1 | - | 2 - 3 | 206 | 71 | ||
BC056388.1 | - | 2 - 3 | 284 | 6854851 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |