Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164173.1 | NP_001157645.1 | 3 - 4 | 421 | 80 | |
NM_053193.2 | NP_444423.1 | 3 - 4 | 421 | 80 | ||
GenBank mRNA | AF322193.1 | - | 3 - 4 | 392 | 80 | |
AK155648.1 | - | 3 - 4 | 362 | 80 | ||
AK155806.1 | - | 3 - 4 | 343 | 80 | ||
BC049941.1 | - | 3 - 4 | 243 | 80 | ||
BC056388.1 | - | 3 - 4 | 421 | 6854851 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |