Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013481.1 | NP_038509.1 | 13 - 14 | 1905 | 78 | |
GenBank mRNA | AK079627.1 | - | 13 - 14 | 1885 | 78 | |
AK129058.1 | - | 13 - 14 | 1878 | 78 | ||
AK144095.1 | - | 13 - 14 | 1917 | 78 | ||
AK167110.1 | - | 13 - 14 | 1888 | 78 | ||
AK167225.1 | - | 13 - 14 | 1889 | 78 | ||
AK167829.1 | - | 13 - 14 | 1883 | 78 | ||
AK167878.1 | - | 13 - 14 | 1893 | 78 | ||
AK168127.1 | - | 13 - 14 | 1886 | 78 | ||
BC012693.1 | - | 13 - 14 | 1879 | 4193123 | 78 | |
U77415.1 | - | 13 - 14 | 1905 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |