Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019536.1 | NP_062409.1 | 77 - 78 | 13565 | 68 | |
GenBank mRNA | AK147443.1 | - | 29 - 30 | 7190 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |