Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195034.1 | NP_001181963.1 | 4 - 5 | 448 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |