Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195034.1 | NP_001181963.1 | 6 - 7 | 641 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |