Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmklr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008153.3 | NP_032179.1 | 2 - 3 | 257 | 71 | |
GenBank mRNA | AK158424.1 | - | 2 - 3 | 174 | 71 | |
BC100612.1 | - | 2 - 3 | 257 | 2654794 | 71 | |
BY347978.1 | - | 2 - 3 | 294 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |