Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145218.3 | NP_660253.2 | 1 - 2 | 1154 | 62 | |
GenBank mRNA | AB175684.1 | - | 1 - 2 | 232 | 62 | |
AF467979.1 | - | 1 - 2 | 441 | 62 | ||
AK035817.1 | - | 1 - 2 | 908 | 62 | ||
AK051281.1 | - | 1 - 2 | 1554 | 62 | ||
AK051382.1 | - | 1 - 2 | 1506 | 62 | ||
AK081848.1 | - | 1 - 2 | 548 | 62 | ||
AK087726.1 | - | 1709 | 62 | |||
BC052469.1 | - | 1 - 2 | 550 | 6810287 | 62 | |
BC069177.1 | - | 1 - 2 | 1154 | 62 | ||
BC158110.1 | - | 1 - 2 | 1181 | 9088184 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |