Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500002828.mm.1 |
NM_199449.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002828.mm.1 |
NM_199449.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002828.mm.1 |
TC1500000404.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500002823.mm.1 |
NM_199449.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002823.mm.1 |
NM_199449.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002823.mm.1 |
TC1500000404.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500002827.mm.1 |
NM_199449.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002827.mm.1 |
NM_199449.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002827.mm.1 |
TC1500000404.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500002824.mm.1 |
NM_199449.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002824.mm.1 |
NM_199449.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002824.mm.1 |
TC1500000404.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500002826.mm.1 |
NM_199449.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002826.mm.1 |
NM_199449.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002826.mm.1 |
TC1500000404.mm.1 |
MTA TC |