Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285517.1 | NP_001272446.1 | 3 - 4 | 348 | 57 | |
NM_001285518.1 | NP_001272447.1 | 3 - 4 | 351 | 57 | ||
NM_001285519.1 | NP_001272448.1 | 3 - 4 | 348 | 57 | ||
NM_009473.3 | NP_033499.1 | 3 - 4 | 355 | 57 | ||
GenBank mRNA | AK054508.1 | - | 3 - 4 | 335 | 57 | |
AK077620.1 | - | 3 - 4 | 330 | 57 | ||
AK134489.1 | - | 3 - 4 | 310 | 57 | ||
AK156777.1 | - | 3 - 4 | 332 | 57 | ||
BC066025.1 | - | 3 - 4 | 306 | 6829527 | 57 | |
BC106148.1 | - | 2 - 3 | 84 | 57 | ||
BQ573162.1 | - | 3 - 4 | 325 | 57 | ||
EU869275.2 | - | 1 - 2 | 73 | 57 | ||
U09419.1 | - | 2 - 3 | 173 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Nuclear Receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |