Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018866.2 | NP_061354.1 | 3 - 4 | 305 | 96 | |
GenBank mRNA | AF030636.1 | - | 3 - 4 | 300 | 95 | |
AF044196.1 | - | 3 - 4 | 305 | 96 | ||
AK156025.1 | - | 3 - 4 | 319 | 96 | ||
AK160241.1 | - | 3 - 4 | 321 | 96 | ||
BC012965.1 | - | 3 - 4 | 304 | 4217539 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |