Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baz1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011714.2 | NP_035844.2 | 9 - 10 | 3199 | 72 | |
GenBank mRNA | AK037737.1 | - | 3 - 4 | 407 | 72 | |
BC141399.1 | - | 9 - 10 | 3102 | 9056012 | 72 | |
BC171975.1 | - | 9 - 10 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |