Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012001.2 | NP_036131.1 | 5 - 6 | 738 | 102 | |
GenBank mRNA | AF071314.1 | - | 5 - 6 | 607 | 102 | |
AK003752.1 | - | 5 - 6 | 600 | 102 | ||
AK009955.1 | - | 5 - 6 | 610 | 102 | ||
AK167833.1 | - | 5 - 6 | 624 | 102 | ||
BC020461.1 | - | 1 - 2 | 1160 | 102 | ||
BC116800.1 | - | 5 - 6 | 592 | 40126119 | 102 | |
BC116826.1 | - | 5 - 6 | 592 | 40126145 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |