Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153391.2 | NP_700440.2 | 14 - 15 | 1539 | 99 | |
GenBank mRNA | AK052303.1 | - | 14 - 15 | 1519 | 99 | |
AK129413.1 | - | 6 - 7 | 628 | 99 | ||
AK147970.1 | - | 14 - 15 | 1539 | 99 | ||
AK166172.1 | - | 14 - 15 | 1505 | 99 | ||
AY029258.1 | - | 14 - 15 | 1609 | 99 | ||
BC060134.1 | - | 14 - 15 | 1533 | 99 | ||
BC082301.1 | - | 13 - 14 | 1419 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |