Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Letm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173012.3 | NP_766600.2 | 4 - 5 | 1046 | 81 | |
GenBank mRNA | AK031563.1 | - | 4 - 5 | 900 | 81 | |
AK050943.1 | - | 4 - 5 | 1212 | 81 | ||
BC055685.1 | - | 4 - 5 | 1046 | 6403034 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |