Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081137.1 | NP_001074606.1 | 4 - 5 | 446 | 107 | |
GenBank mRNA | EU937530.1 | - | 5 - 6 | 509 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |