Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK076605.1 | - | 13 - 14 | 1712 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |