Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272057.1 | NP_001258986.1 | 13 - 14 | 1690 | 78 | |
NM_001272058.1 | NP_001258987.1 | 13 - 14 | 1690 | 78 | ||
NM_001272059.1 | NP_001258988.1 | 13 - 14 | 1690 | 78 | ||
NM_007401.3 | NP_031427.2 | 13 - 14 | 1690 | 78 | ||
GenBank mRNA | AK016574.1 | - | 13 - 14 | 1346 | 78 | |
AK029519.1 | - | 13 - 14 | 1380 | 78 | ||
AK161262.1 | - | 13 - 14 | 1690 | 78 | ||
AK161415.1 | - | 10 - 11 | 1068 | 78 | ||
AK161417.1 | - | 13 - 14 | 1360 | 78 | ||
BC094237.1 | - | 13 - 14 | 1387 | 30604517 | 78 | |
KF144382.1 | - | 1 - 2 | 91 | 78 | ||
U06150.1 | - | 1 - 2 | 100 | 78 | ||
U22059.1 | - | 13 - 14 | 1354 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |