Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025691.1 | NP_079967.1 | 16 - 17 | 1661 | 144 | |
GenBank mRNA | AK017869.1 | - | 2 - 3 | 144 | ||
BC089462.1 | - | 7 - 8 | 669 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |