Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300003791.mm.1 |
NM_013928.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003791.mm.1 |
NM_013928.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003791.mm.1 |
TC0300003210.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300003794.mm.1 |
NM_001113420.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003794.mm.1 |
NM_001113420.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003794.mm.1 |
NM_001113421.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003794.mm.1 |
NM_001113421.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003794.mm.1 |
NM_001282044.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003794.mm.1 |
NM_001282045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003794.mm.1 |
NM_013928.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003794.mm.1 |
NM_013928.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003794.mm.1 |
TC0300003210.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300003793.mm.1 |
NM_001113420.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003793.mm.1 |
NM_001113420.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003793.mm.1 |
NM_001113421.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003793.mm.1 |
NM_001113421.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003793.mm.1 |
NM_001282044.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003793.mm.1 |
NM_001282045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003793.mm.1 |
NM_013928.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003793.mm.1 |
NM_013928.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300003793.mm.1 |
TC0300003210.mm.1 |
MTA TC |