Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010568.2 | NP_034698.2 | 15 - 16 | 3465 | 64 | |
GenBank mRNA | J05149.1 | - | 15 - 16 | 3024 | 64 | |
KC356871.1 | - | 16 - 17 | 3012 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Mus musculus Probe spans exons |