Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010568.2 | NP_034698.2 | 19 - 20 | 4121 | 75 | |
GenBank mRNA | AK164267.1 | - | 1 - 2 | 117 | 75 | |
KC356871.1 | - | 20 - 21 | 3668 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mus musculus Probe spans exons |