Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042557.2 | NP_001036022.1 | 6 - 7 | 789 | 95 | |
NM_001164172.1 | NP_001157644.1 | 6 - 7 | 789 | 95 | ||
NM_001291777.1 | NP_001278706.1 | 5 - 6 | 741 | 95 | ||
NM_001291778.1 | NP_001278707.1 | 5 - 6 | 632 | 95 | ||
NM_001291783.1 | NP_001278712.1 | 5 - 6 | 632 | 95 | ||
NM_011944.3 | NP_036074.2 | 5 - 6 | 741 | 95 | ||
GenBank mRNA | AB005654.1 | - | 6 - 7 | 664 | 95 | |
AF022112.1 | - | 5 - 6 | 718 | 95 | ||
AF022113.1 | - | 6 - 7 | 766 | 95 | ||
AF060943.1 | - | 5 - 6 | 632 | 95 | ||
AF060944.1 | - | 5 - 6 | 652 | 95 | ||
AF060945.1 | - | 5 - 6 | 652 | 95 | ||
AF060946.1 | - | 6 - 7 | 700 | 95 | ||
AF060947.1 | - | 6 - 7 | 700 | 95 | ||
AK031137.1 | - | 5 - 6 | 650 | 95 | ||
AK165184.1 | - | 5 - 6 | 658 | 95 | ||
BC070467.1 | - | 6 - 7 | 701 | 30615302 | 95 | |
U74463.1 | - | 5 - 6 | 598 | 95 | ||
U74464.1 | - | 4 - 5 | 487 | 95 | ||
U93030.1 | - | 5 - 6 | 632 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |