Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029465.3 | NP_083741.1 | 6 - 7 | 480 | 70 | |
GenBank mRNA | AK079992.1 | - | 6 - 7 | 481 | 70 | |
BC025069.1 | - | 6 - 7 | 472 | 70 | ||
BC087963.1 | - | 6 - 7 | 480 | 6766377 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |