Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026156.2 | NP_080432.1 | 1 - 2 | 74 | 98 | |
GenBank mRNA | AK002890.1 | - | 1 - 2 | 75 | 98 | |
AK031533.1 | - | 84 | 98 | |||
AK087952.1 | - | 1 - 2 | 81 | 98 | ||
AK132434.1 | - | 1 - 2 | 88 | 98 | ||
BC021341.1 | - | 1 - 2 | 72 | 5066146 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |