Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hook3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207659.3 | NP_997542.1 | 5 - 6 | 611 | 73 | |
GenBank mRNA | AK054263.1 | - | 5 - 6 | 530 | 73 | |
AK080377.1 | - | 5 - 6 | 578 | 73 | ||
AK145149.1 | - | 5 - 6 | 581 | 73 | ||
AK170858.1 | - | 5 - 6 | 558 | 73 | ||
AY223806.1 | - | 5 - 6 | 401 | 73 | ||
BC090960.1 | - | 5 - 6 | 521 | 30651062 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |