Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF099067.1 | - | 13 - 14 | 2289 | 115 | |
AJ223071.1 | - | 13 - 14 | 2379 | 115 | ||
AK078809.1 | - | 12 - 13 | 2256 | 115 | ||
AK132803.1 | - | 12 - 13 | 2173 | 115 | ||
AK149147.1 | - | 4 - 5 | 457 | 115 | ||
AK149584.1 | - | 12 - 13 | 2173 | 115 | ||
BC057939.1 | - | 13 - 14 | 2291 | 5325558 | 115 | |
Y09234.1 | - | 12 - 13 | 2235 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |