Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011103.3 | NP_035233.1 | 6 - 7 | 809 | 77 | |
GenBank mRNA | AB011812.1 | - | 5 - 6 | 572 | 77 | |
AB201453.1 | - | 3 - 4 | 411 | 77 | ||
AB201454.1 | - | 3 - 4 | 411 | 77 | ||
AB201455.1 | - | 3 - 4 | 411 | 77 | ||
AB201456.1 | - | 3 - 4 | 411 | 77 | ||
AF251036.1 | - | 5 - 6 | 572 | 77 | ||
AK144231.1 | - | 6 - 7 | 814 | 77 | ||
AK168768.1 | - | 6 - 7 | 804 | 77 | ||
AK171476.1 | - | 6 - 7 | 811 | 77 | ||
AY545076.1 | - | 5 - 6 | 572 | 77 | ||
BC051416.1 | - | 6 - 7 | 773 | 6515302 | 77 | |
M69042.1 | - | 6 - 7 | 804 | 77 | ||
X60304.1 | - | 5 - 6 | 585 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |