Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500005709.mm.1 |
TC0500000647.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500005709.mm.1 |
NM_001285452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005709.mm.1 |
NM_028975.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005709.mm.1 |
NM_030108.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005715.mm.1 |
TC0500000647.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500005715.mm.1 |
NM_001285452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005715.mm.1 |
NM_028975.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005715.mm.1 |
NM_030108.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005704.mm.1 |
TC0500000647.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500005704.mm.1 |
NM_001285452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005704.mm.1 |
NM_028975.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005704.mm.1 |
NM_030108.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005708.mm.1 |
TC0500000647.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500005708.mm.1 |
NM_001285452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005708.mm.1 |
NM_028975.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005708.mm.1 |
NM_030108.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005705.mm.1 |
TC0500000647.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500005705.mm.1 |
NM_001285452.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005705.mm.1 |
NM_028975.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500005705.mm.1 |
NM_030108.2 |
Refseq |