Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161373.1 | NP_001154845.1 | 10 - 11 | 1938 | 78 | |
NM_001161374.1 | NP_001154846.1 | 10 - 11 | 1938 | 78 | ||
NM_007977.2 | NP_032003.2 | 10 - 11 | 1938 | 78 | ||
GenBank mRNA | BC138347.1 | - | 10 - 11 | 1819 | 8861367 | 78 |
BC145245.1 | - | 10 - 11 | 1819 | 9053797 | 78 | |
L05573.1 | - | 10 - 11 | 1936 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |