Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tacc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040435.2 | NP_001035525.1 | 13 - 14 | 2285 | 64 | |
GenBank mRNA | AF093542.1 | - | 14 - 15 | 1519 | 64 | |
AF156934.1 | - | 13 - 14 | 1839 | 64 | ||
AF203445.1 | - | 13 - 14 | 1786 | 64 | ||
AF203446.1 | - | 13 - 14 | 1174 | 64 | ||
AF247674.1 | - | 14 - 15 | 1570 | 64 | ||
AK076282.1 | - | 2312 | 64 | |||
AK145556.1 | - | 13 - 14 | 1846 | 64 | ||
AK160291.1 | - | 13 - 14 | 1848 | 64 | ||
AK179770.1 | - | 2 - 3 | 135 | 64 | ||
BC003252.1 | - | 13 - 14 | 1842 | 3500413 | 64 | |
BC055071.1 | - | 4261 | 64 | |||
BC066184.1 | - | 13 - 14 | 1821 | 30457053 | 64 | |
BC068314.1 | - | 13 - 14 | 1821 | 30461783 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |