Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpeb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175937.3 | NP_787951.2 | 3 - 4 | 1997 | 107 | |
GenBank mRNA | AB100307.1 | - | 4 - 5 | 630 | 107 | |
BC107349.1 | - | 4 - 5 | 607 | 40055453 | 107 | |
BC107350.1 | - | 4 - 5 | 607 | 40055458 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |