Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpeb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177379.1 | NP_001170850.1 | 8 - 9 | 2457 | 85 | |
NM_175937.3 | NP_787951.2 | 8 - 9 | 2523 | 85 | ||
GenBank mRNA | AB100307.1 | - | 9 - 10 | 1156 | 85 | |
AK042065.1 | - | 9 - 10 | 1094 | 85 | ||
AK076221.1 | - | 2 - 3 | 219 | 85 | ||
BC107349.1 | - | 9 - 10 | 1133 | 40055453 | 85 | |
BC107350.1 | - | 9 - 10 | 1133 | 40055458 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |