Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Letm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019694.1 | NP_062668.1 | 10 - 11 | 1751 | 76 | |
GenBank mRNA | AB041582.1 | - | 10 - 11 | 1764 | 76 | |
AF061026.1 | - | 10 - 11 | 1751 | 76 | ||
AK133849.1 | - | 10 - 11 | 1771 | 76 | ||
AK161638.1 | - | 4 - 5 | 1473 | 76 | ||
BC023862.1 | - | 10 - 11 | 1748 | 76 | ||
BC046326.1 | - | 10 - 11 | 1758 | 76 | ||
BC055865.1 | - | 10 - 11 | 1769 | 76 | ||
BC061115.1 | - | 10 - 11 | 1764 | 6533034 | 76 | |
BC061690.1 | - | 10 - 11 | 1751 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |