Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fndc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022424.5 | NP_071869.2 | 1 - 2 | 343 | 112 | |
GenBank mRNA | AK013203.1 | - | 1 - 2 | 337 | 112 | |
AK018154.1 | - | 1 - 2 | 339 | 112 | ||
AK163183.1 | - | 1 - 2 | 344 | 112 | ||
BC139278.1 | - | 1 - 2 | 95 | 8862300 | 112 | |
BC139292.1 | - | 1 - 2 | 95 | 8862314 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |