Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028102.1 | NP_082378.1 | 7 - 8 | 956 | 71 | |
GenBank mRNA | AK035532.1 | - | 7 - 8 | 1007 | 71 | |
AK079311.1 | - | 7 - 8 | 1004 | 71 | ||
BC046229.1 | - | 6 - 7 | 876 | 71 | ||
BC118962.2 | - | 7 - 8 | 1192 | 40095026 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |