Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009619.4 | NP_033749.2 | 19 - 20 | 82 | ||
GenBank mRNA | BC049609.1 | - | 7 - 8 | 82 | ||
BC129920.1 | - | 19 - 20 | 2246 | 82 | ||
X64227.1 | - | 19 - 20 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |