Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008439.3 | NP_032465.2 | 5 - 6 | 582 | 63 | |
GenBank mRNA | AK008326.1 | - | 1 - 2 | 125 | 63 | |
AK008503.1 | - | 1 - 2 | 125 | 63 | ||
BC013464.1 | - | 5 - 6 | 582 | 4235560 | 63 | |
Y09335.1 | - | 5 - 6 | 561 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |