Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sema4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163489.1 | NP_001156961.1 | 9 - 10 | 1052 | 78 | |
NM_001163490.1 | NP_001156962.1 | 9 - 10 | 1131 | 78 | ||
NM_001163491.1 | NP_001156963.1 | 9 - 10 | 1090 | 78 | ||
NM_013658.3 | NP_038686.3 | 9 - 10 | 1230 | 78 | ||
GenBank mRNA | AK035918.1 | - | 8 - 9 | 1092 | 78 | |
AK077976.1 | - | 9 - 10 | 1210 | 78 | ||
AK152728.1 | - | 9 - 10 | 1222 | 78 | ||
AK153164.1 | - | 9 - 10 | 1222 | 78 | ||
AK154392.1 | - | 9 - 10 | 1191 | 78 | ||
BC025800.1 | - | 9 - 10 | 1148 | 5102015 | 78 | |
X85991.1 | - | 9 - 10 | 1052 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |