Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081001.2 | NP_001074470.1 | 17 - 18 | 8249 | 67 | |
NM_009765.3 | NP_033895.2 | 17 - 18 | 8055 | 67 | ||
GenBank mRNA | AK165522.1 | - | 7 - 8 | 1384 | 67 | |
BC059904.1 | - | 7 - 8 | 1512 | 67 | ||
U65594.1 | - | 17 - 18 | 8032 | 67 | ||
U82270.1 | - | 16 - 17 | 7741 | 67 | ||
U89652.1 | - | 16 - 17 | 7779 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |