Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009765.3 | NP_033895.2 | 1 - 2 | 274 | 88 | |
GenBank mRNA | AK148272.1 | - | 1 - 2 | 272 | 88 | |
AK164598.1 | - | 1 - 2 | 264 | 88 | ||
U65594.1 | - | 1 - 2 | 251 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus 1700 Probe spans exons |