Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081001.2 | NP_001074470.1 | 24 - 25 | 9523 | 80 | |
NM_009765.3 | NP_033895.2 | 24 - 25 | 9329 | 80 | ||
GenBank mRNA | BC059904.1 | - | 14 - 15 | 2786 | 80 | |
U65594.1 | - | 24 - 25 | 9306 | 80 | ||
U82270.1 | - | 23 - 24 | 9015 | 80 | ||
U89652.1 | - | 23 - 24 | 9053 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |