Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slmap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032008.3 | NP_114397.3 | 7 - 8 | 902 | 122 | |
GenBank mRNA | AK052158.1 | - | 8 - 9 | 1868 | 122 | |
AK129403.1 | - | 7 - 8 | 902 | 122 | ||
AK141597.1 | - | 7 - 8 | 2059 | 122 | ||
AK163630.1 | - | 8 - 9 | 1841 | 122 | ||
AK164911.1 | - | 7 - 8 | 2057 | 122 | ||
AK166396.1 | - | 7 - 8 | 2058 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |