Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146097.3 | NP_666209.1 | 8 - 9 | 741 | 143 | |
NR_033744.1 | 8 - 9 | 808 | 143 | |||
GenBank mRNA | AK076027.1 | - | 8 - 9 | 696 | 143 | |
AK132291.1 | - | 8 - 9 | 727 | 143 | ||
BC018472.1 | - | 8 - 9 | 683 | 3661842 | 143 | |
BC053745.1 | - | 8 - 9 | 776 | 6505520 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |